Cleanroom hoods, for use with goggles, 5R181
Dodavatel: FRISTADS KANSAS
Certifikáty
O tomto produktu
These cleanroom hoods are made of XR50 fabric, consisting of 97% polyester and 3% conductive fibre. It is ideal for sterile environments. The fabric has no change in pore size after 50 wash and autoclave cycles.
- Face opening suitable for use of goggles
- Adjustable with tie tape
- Full shoulder cover
Weight: 115 g/m²
Specifikace
- Catalog No:
- KWIN100649500
- KWIN100649730
- Model:
- 5R181
- 5R181
- Aplikace oblečení:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Rod:
- Unisex
- Unisex
- Tkanina:
- XR50 fabric (97% polyester and 3% conductive fibre)
- XR50 fabric (97% polyester and 3% conductive fibre)
- Hmotnost:
- Weight: 115 g/m²
- Weight: 115 g/m²