Orders LinkContactUsLinkComponent
Teaching camera
Digital teaching camera, 3 MP
Katalogové číslo DIDAPOF010810
Dodavatel:   DIDALAB
Teaching camera
Katalogové číslo DIDAPOF010810
Dodavatel:  DIDALAB
Teaching camera
Teaching camera
Digital teaching camera, 3 MP
Přejít na:
Price
28 380,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
The pedagogical model of the digital camera can be put on a Ø-10 mm rod or on a camera’s stand.
  • Pentax lenses: One with a fixed 50-mm focus, the other with a 28 to 70-mm focus
  • Focus, aperture, time exposure, field depth, magnification, sensor, sensibility and accuracy
  • Image reflection with mirror to understand the Reflex aiming
By setting the lens, the students can understand the field depth and the digital aperture principle.
Delivery information: Supplied with specific software and a complete manual.
Specifikace
  • Rozlišení:
    3 MP
  • Popis:
    Digital teaching camera