High visibility trousers, bi-coloured, Portwest Texo, TX51
: Portwest
These modern trousers with contrast colouring are made of 65% polyester and 35% cotton fabric which offers comfort and durability. Compatible with the TX50 jacket.
- Mobile phone pocket, pen pockets
- Kneepad pockets
- Adjustable Velcro hems
- Back elasticated waistband
Weight: 245 g/m²
: EN ISO 20471 (Class 2)
- Catalog No:
- PRTWTX51YNRS
- PRTWTX51YNRM
- PRTWTX51YNRL
- PRTWTX51ONRXXL
- PRTWTX51ONRXL
- PRTWTX51ONRS
- PRTWTX51ONRM
- PRTWTX51ONRL
- PRTWTX51YNRXXL
- PRTWTX51YNRXL
- Model:
- TX51
- TX51
- TX51
- TX51
- TX51
- TX51
- TX51
- TX51
- TX51
- TX51
- Aplikace oblečení:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Rod:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tkanina:
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- Hmotnost:
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
Frequently Bought Together