High visibility softshell jacket, removable sleeves, S428
Dodavatel: Portwest
Certifikáty
O tomto produktu
Wind- and water-resistant jacket made of 94% polyester and 6% Spandex laminated to a stretchable polyester fleece fabric.
- Inner fabric is a bonded micro polar fleece for added warmth
- With zip-off sleeves
- All zips are waterproof
- Multiple pockets provide ample storage
Weight: 310 g/m²
Certifications: EN ISO 40271 (Class 3 with sleeves, class 2 without sleeves)
Specifikace
- Catalog No:
- PRTWS428YERXXXL
- PRTWS428YERXL
- PRTWS428YERXXL
- PRTWS428YERS
- PRTWS428YERM
- PRTWS428YERL
- PRTWS428ORRS
- PRTWS428ORRL
- PRTWS428ORRM
- PRTWS428ORRXL
- PRTWS428ORRXXL
- PRTWS428ORRXXXL
- PRTWS428YER4XL
- PRTWS428ORR4XL
- Model:
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- S428
- Aplikace oblečení:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rod:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Typ uzávěru:
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Se zipem
- Tkanina:
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- 94% polyester and 6% Spandex
- Hmotnost:
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²
- 310 g/m²