VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 9 Elements in Hydrochloric Acid 10% for ICP
Catalog # 84792.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Matrice:10% HCl
- Typ balení:Plastic bottle
- Bal.:100 mL
- Popis:Multi-element quality control standard 100 mg/l {Au; Ir; Pd; Pt; Rh; Ru; Os; Sn; Te} ARISTAR® pro ICP
Specifications
About this item
9 Elements 100 mg/L: Au, Ir, Pd, Pt, Rh, Ru, Os, Sn, Te.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.