Closed top caps for storage vials
Katalogové číslo NSCAB7815-20
Dodavatel: THERMO NATIONAL SCIENTIFIC
Specifikace
- Septum:PTFE
- Popis:Closed top cap, 20-400, PP
- Sekvence:05
- Bal.:100
Specifikace
O tomto produktu
PTFE on film/foam backing offers broad chemical resistance.
Not autoclavable.