Bottles, wide neck, round, with screw cap, Precisionware®, SP Bel-Art
Katalogové číslo BELAF106320008
Dodavatel: Bel-Art Products, a Part of SP
Specifikace
- Závit:53 mm
- Type:with heavy knurled PP caps
- Kapacita:1000 ml
- Bal.:6
Specifikace
O tomto produktu
For pharmaceutical or similar packaging of liquid or dry materials. Polypropylene bottles are Boston-round style, clear, and moulded from FDA compliant material. Polypropylene caps have a valve-seal design for a leakproof fit. Deep, square threads help prevent back-off during shipping. Ribbed cap exteriors are designed for torque closure and easy opening. Smooth, rounded bottle interiors are designed for easy cleaning and thorough emptying. Autoclavable, with caps detached, to 121 °C (250 °F).