Filtering flasks, heavy wall with tubulation, PYREX®
Catalog # CORN5340-2L
Supplier: Corning
Specifications
- Ø vněj.:168 mm
- Kapacita:2000 ml
- Výška:292 mm
- Sekvence:04
- Bal.:1
Specifications
About this item
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.
The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.