ContactUsLinkComponent
Screw cap with rubber liner, reusable, Corning®
Screw cap with rubber liner, reusable, Corning®
Catalog # CORN9999-241
Supplier:  Corning
   Low Manufacturing Impact
undefined
Screw cap with rubber liner, reusable, Corning®
Catalog # CORN9999-241
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Závit:
    24-410
  • Barva:
    Černá
  • Materiál:
    Fenol
  • Sekvence:
    01
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Bal.:
    144
  • Certifikát ochrany životního prostředí:
    I-RECs, ISO 14001 - Monterrey