Cesium chloride-aluminium nitrate buffer solution acc. to Schuhknecht and Schinkel, Supelco®
Catalog # 1.02037.1000
Supplier: Merck
CAS Number:
Specifications
- Bal.:1 L
- Popis:Cesium chloride-aluminium nitrate buffer solution acc. to Schuhknecht and Schinkel for the determination of sodium, potassium and lithium by flame spectrometry (enth./cont. 50 g/l CsCl + 250 g/l Al(NO₃)₃ * 9 H₂O)
- M-Clarity quality level:MQ100