Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Isoquinolineboronic acid
4-Isoquinolineboronic acid 97%
4-Isoquinolineboronic acid 97%
4-Isoquinolineboronic acid 97%
ABCRAB172568.0001
:
abcr
:
4-Isoquinolineboronic acid 97%
ABCRAB172568.0001
:
abcr
:
Typ balení:
Glass bottle for solids
Bal.:
1 g
Sekvence:
3047
:
:
172.9763 g/mol
:
>270 °C
:
Freezer
:
192182-56-2
:
C₉H₈BNO₂
:
MFCD03412093