

Chloroform
Catalog # LACH31058
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1 L
- Sekvence:5037
Specifications
About this item
:
Merck Index:13,02160
Boiling Pt:61,7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1,4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
REACH:05-2114614906-45
:GHS06
:GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826