bromfenolová modř
Catalog # LACH40097IP0G0010
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:10 g
- Sekvence:3236
Specifications
About this item
:
Merck Index:13,01429
Melting Pt:273 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-086-2
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875