Hydrogensíran draselný
4024, Plastová láhev, 1 kg
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Potassium bisulphate
- CAS Number:7646-93-7
- Formula:KHSO₄
- MDL Number:MFCD00011404
- Molecular Weight:136.17
- Merck Index:13,07695
- Melting Pt:214 °C
- Density:2,32 g/cm³ (20 °C)
- EINECS:231-594-1
- UN:2509
- ADR:8,II
- :Density: 2,32 g/cm³ (20 °C)
Documents
Specifications
Specifications
- Typ balení:Plastová láhev
- Bal.:1 kg
- Sekvence:4024



