Síran železnato-amonný hexahydrát
Supplier: Lach-ner S.R.O.
Certificates
About this item
Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
CAS Number:7783-85-9
:
Formula:(NH₄)₂Fe(SO₄)₂·6H₂O
MDL Number:MFCD00150530
Melting Pt:100 °C
Density:1,864 g/cm³ (20 °C)
EINECS:233-151-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together