Separating funnel, globe shaped
Catalog # KAVA426351940
Supplier: Kavalier
Specifications
- Velikost objímky:29/32 NS
- Kapacita:1000 ml
- Sekvence:02
- Bal.:1
Specifications
About this item
Borosilicate glass 3.3.
- Very good chemical resistance
- Very good temperature resistance
Suitable for separation of particular phases of the mixture. With standard ground joint stopper.