Flasks, flat bottom with standard ground joint
Catalog # KAVA632426217250
Supplier: Kavalier
Specifications
- Ø vněj.:85 mm
- Velikost objímky:29/32 NS
- Kapacita:250 ml
- Výška:140 mm
- Bal.:1
Specifications
About this item
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.