Ball Valve for Storage Bottles
Catalog # 229-7489
Supplier: SEMADENI
Specifications
- Velikost portu:½" NW 16
- Popis:Ball valve, PP, white
- Bal.:1
Specifications
About this item
½” universal valve made from PP, resistant to acids, alkalis and solvents.
- Fully sealed design, runs very easily and with no blockages
- Outlet spout Ø 12 mm
Housing and ball made from PP, ball seal made from PTFE and O ring made from nitrile rubber (NBR 70).