Order Entry
Czech Republic
ContactUsLinkComponent
Cyklohexan
Flammable
Health
Irritant
Environment
Cyklohexan
Catalog # LACH30106
Supplier:  Lach-ner S.R.O.
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Cyklohexan
Catalog # LACH30106
Supplier:  Lach-ner S.R.O.
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    2,5 L
  • Sekvence:
    5096

Specifications

About this item

 :
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0,7785 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814