Orders LinkContactUsLinkComponent
Tetrachlorethylen
5037, Glass bottle, 1 L
  LACH20055CT0M1000
Tetrachlorethylen
  LACH20055CT0M1000
 :  127-18-4
Tetrachlorethylen
Tetrachlorethylen
5037, Glass bottle, 1 L
 :  127-18-4
390,00 Kč
1 * 1 L
 :  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  •  :127-18-4
  •  :C₂Cl₄
  •  :MFCD00000834
  •  :165.83
  •  :12,09332
  •  :121,1 °C (1013 hPa)
  •  :–22 °C
  •  :1,63 g/cm³ (20 °C)
  •  :Ambient
  •  :204-825-9
  •  :6.1,III
  •  :Density: 1,63 g/cm³ (20 °C)
  •  :1897
  • Typ balení:
    Glass bottle
  • Bal.:
    1 L
  • Sekvence:
    5037