n-Hexan
5037, Glass bottle, 1 L
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94,3 °C
- Density:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- UN:1208
- ADR:3,II
- :Density: 0,659 g/cm³ (20 °C)
- EINECS:203-777-6
Documents
Specifications
Specifications
- Typ balení:Glass bottle
- Bal.:1 L
- Sekvence:5037



