n-Hexan
5096, Glass bottle, 2,5 L
Catalog # LACH20032R99M2500
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94,3 °C
- Density:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- UN:1208
- ADR:3,II
- :Density: 0,659 g/cm³ (20 °C)
- EINECS:203-777-6
Specifications
Specifications
- Typ balení:Glass bottle
- Bal.:2,5 L
- Sekvence:5096
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



