1-Propanol
5096, Glass bottle, 2,5 L
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
- CAS Number:71-23-8
- Formula:H₃CCH₂CH₂OH
- MDL Number:MFCD00002941
- Molecular Weight:60.1
- Merck Index:13,07934
- Boiling Pt:97 °C (1013 hPa)
- Melting Pt:–127 °C
- Density:0,8053 g/cm³ (20 °C)
- Flash Pt:15 °C
- Storage Temperature:Ambient
- UN:1274
- ADR:3,II
- :Density: 0,8053 g/cm³ (20 °C)
- EINECS:200-746-9
Documents
Specifications
Specifications
- Typ balení:Glass bottle
- Bal.:2,5 L
- Sekvence:5096
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



