


1-Butanol
Catalog # LACH20010CT0M1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1 L
- Sekvence:5037
Specifications
About this item
:
Merck Index:13,01539
Boiling Pt:117,6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,812 g/cm³ (20 °C)
Flash Pt:30 °C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
:GHS02
:GHS05
:GHS07
CAS Number:71-36-3
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964