Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Dichlormethan

Dichlormethan

Health
Irritant
Supplier:   Lach-ner S.R.O.
Dichlormethan
Supplier:  Lach-ner S.R.O.
Certificates
Dichlormethan

3 Options Available Below

Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93
  • Merck Index:12,06140
  • Boiling Pt:39,8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1,322 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Density: 1,322 g/cm³ (20 °C)
  • UN:1593
Product Family Options

Product Information

  • Sekvence
    Typ balení
    Bal.
    Availability
    Price
  • 5037
    Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 377,00 Kč
    377,00 Kč
    1 * 1 L
       377,00 Kč

  • 5096
    Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 938,00 Kč
    938,00 Kč
    1 * 2,5 L
       938,00 Kč

  • 5190
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 1 350,00 Kč
    1 350,00 Kč
    1 * 5 L
       1 350,00 Kč