
Kyselina octová ≥80%
Catalog # LACH10047A80M1000
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:1 L
- Sekvence:5037
Specifications
About this item
:
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152