N,N-Dimethylformamid
5096, Glass bottle, 2,5 L
LACH200211T0M25003
: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- :68-12-2
- :HCON(CH₃)₂
- :MFCD00003284
- :73.09
- :13,03269
- :153 °C (1013 hPa)
- :–61 °C
- :0,949 g/cm³ (20 °C)
- :57,5 °C (ot. pohárek)
- :2265
- :3,III
- :Density: 0,949 g/cm³ (20 °C)
- :Ambient
- :200-679-5
- :01-2119475605-32
- Typ balení:Glass bottle
- Bal.:2,5 L
- Sekvence:5096