Orders LinkContactUsLinkComponent
N,N-Dimethylformamid
5096, Glass bottle, 2,5 L
  LACH200211T0M25003
N,N-Dimethylformamid
  LACH200211T0M25003
 :  68-12-2
N,N-Dimethylformamid
N,N-Dimethylformamid
5096, Glass bottle, 2,5 L
 :  68-12-2
1 * 2,5 L
 :  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  •  :68-12-2
  •  :HCON(CH₃)₂
  •  :MFCD00003284
  •  :73.09
  •  :13,03269
  •  :153 °C (1013 hPa)
  •  :–61 °C
  •  :0,949 g/cm³ (20 °C)
  •  :57,5 °C (ot. pohárek)
  •  :2265
  •  :3,III
  •  :Density: 0,949 g/cm³ (20 °C)
  •  :Ambient
  •  :200-679-5
  •  :01-2119475605-32
  • Typ balení:
    Glass bottle
  • Bal.:
    2,5 L
  • Sekvence:
    5096