Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Hydraziniumsulfát

Hydraziniumsulfát

Toxic
Health
Environment
Supplier:   Lach-ner S.R.O.
Hydraziniumsulfát
Supplier:  Lach-ner S.R.O.
Certificates
Hydraziniumsulfát
Hydraziniumsulfát

3 Options Available Below

Product Details & Documents
Synonyms:  Hydrazine sulphate salt, Hydrazine sulphate
  • CAS Number:10034-93-2
  • Formula:NH₂NH₂·H₂SO₄
  • MDL Number:MFCD00044873
  • Molecular Weight:130.12
  • Merck Index:14,04772
  • Melting Pt:254 °C
  • Density:1,37 g/cm³ (20 °C)
  • EINECS:233-110-4
  • UN:3288
  • ADR:6.1,III
  •  :Density: 1,37 g/cm³ (20 °C)
Product Family Options

Product Information

  • Sekvence
    Typ balení
    Bal.
    Availability
    Price
  • 3401
    Plastová láhev
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

  • 3545
    Plastová láhev
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

  • 3645
    Plastová láhev
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.