Fenolová červeň
Catalog # LACH30114
Supplier: Lach-ner S.R.O.
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:25 g
- Sekvence:3308
Specifications
About this item
:
Merck Index:13,07329
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:205-609-7
CAS Number:143-74-8
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552