Copper(II) nitrate
3645, Plastová láhev, 500 g
Catalog # LACH30036CP0G0500
Supplier: Lach-ner S.R.O.
Jump to:
Product Details & Documents
Synonyms: Cupric nitrate, Copper dinitrate
- CAS Number:3251-23-8
- Formula:CuN₂O₆
- Molecular Weight:187.56
- Merck Index:12,02710
- Melting Pt:114,5 °C
- Density:2,32 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:221-838-5
- UN:1477
- ADR:5.1,II
- :Density: 2,32 g/cm³ (20 °C)
Specifications
Specifications
- Typ balení:Plastová láhev
- Bal.:500 g
- Sekvence:3645
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



