Orders LinkContactUsLinkComponent
Assortment Boxes
with 2×6 compartments, 250×170×45 mm
Katalogové číslo SEMA0457
Dodavatel:   SEMADENI
Assortment Boxes
Katalogové číslo SEMA0457
Dodavatel:  SEMADENI
Assortment Boxes
Assortment Boxes
with 2×6 compartments, 250×170×45 mm
Přejít na:
Price
882,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Assortment and bearing boxes, made of PS, transparent, with hinged lid and simple longitudinal and transverse divisions.
Specifikace
  • Popis:
    with 2×6 compartments
  • Š×H×V:
    250×170×45 mm