Zirconium standard dissolved in oil, 5.000 µg/g Zr in mineral oil Specpure® pro wear metal analysis
Dodavatel: Thermo Fisher Scientific
Certifikáty
O tomto produktu
Physical form: liquid
MDL No.: MFCD00011303
Specifikace
- MDL čí.:MFCD00011303