Sample loops for Rheodyne® sample injectors
Catalog # HYPE9055-022
Supplier: Thermo Fisher Scientific
Specifications
- Kapacita:20 µl
- Popis:Sample loop, for 9010, 9725 and 9725i injectors
- Materiál:PEEK
- Sekvence:08
- Bal.:1
- Pro zkumavky s vnitř. Ø:0,25 mm