Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Filtry
Jednotky filtrační membránové
Accessories for online filtration system MicropreSure™
Accessories for online filtration system MicropreSure™
Accessories for online filtration system MicropreSure™
MILXM00000001
:
Merck
Accessories for online filtration system MicropreSure™
MILXM00000001
:
Merck
Popis:
Sterile sampling tube 5 in. w/Luer fitting
Sekvence:
14
Bal.:
100