Microarray hybridisation solution v2.0
Catalog # RPK0325
Supplier: Cytiva
CAS Number:
Specifications
- Bal.:1
- Popis:Microarray Hybridisation Solution v2.0
Specifications
About this item
Microarray hybridisation solution version 2.0, for highly reproducible microarray results.
- Includes a blend of proprietary components to provide highly reproducible, high-quality automated and manual DNA microarray hybridisations
- Hybridisation rate enhancers improve the detection of rare targets
- Proprietary blocking agents minimize non-specific background
DNA microarrays can be used to measure the expression levels of large numbers of genes simultaneously (gene expression profiling), or to genotype multiple areas of a genome (SNP arrays), with variable coverage or resolution depending how many DNA 'spots' cover those areas.