Absorbent mat pads for viscous liquids, PIG®
Catalog # NEWPMAT209
Supplier: NEW PIG
Specifications
- Popis:Heavy fluids mat pads
- D׊:560×460 mm
- Savost:182 litres/bag
- Bal.:50
Specifications
About this item
White polyester fibre absorbent mat pads for viscous liquids for picking up paint, glue, resins, shampoo, liquid chocolate, syrup, varnish.
Up to 3,63 litres of viscous fluids per pad.