Orders LinkContactUsLinkComponent
Nucleic acid labelling and detection system, AlkPhos Direct™
Nucleic acid labelling and detection system, AlkPhos Direct™
Dodavatel:   Cytiva
Nucleic acid labelling and detection system, AlkPhos Direct™
Dodavatel:  Cytiva
Certifikáty
Nucleic acid labelling and detection system, AlkPhos Direct™
Nucleic acid labelling and detection system, AlkPhos Direct™
3 Dostupných variant níže
Product Details & Documents
AlkPhos Direct™ Labelling and detection systems are based on the rapid, direct labelling of DNA or RNA probes with thermostable alkaline phosphatase. AlkPhos Direct™ combines the convenience of direct enzyme labelling (no blocking or antibody stages) with those of alkaline phosphatase detection (long light output and high sensitivity).
  • Fast and convenient system for all high sensitivity applications using DNA, RNA, or oligonucleotide probes
  • Hybridisation stringency can be controlled by elevating temperatures up to 75 °C
  • Detection of 60 fg target DNA (single-copy gene) in a genomic southern blot
  • ECF detection for quantitation experiments
  • Saves 2 to 3 hours over conventional indirect methods
Labelling is complete within 30 minutes in a single tube protocol and the resulting probe is ready for use in hybridisations without further purification. Due to the thermostable nature of the enzyme, hybridisation stringency can be controlled by adjusting temperature as well as salt concentration. The system can be used with either chemiluminescent or fluorescent detection reagents. The most sensitive result is achieved with CDP-Star™, a chemiluminescent substrate with a fast light output. Alternatively, the use of ECF substrate generates a fluorescent signal suitable for use with fluorescence scanning devices.
Informace o dodávce: Each system includes labelling reagent, crosslinker, control DNA, reaction buffer, hybridisation buffer, blocking agent, CDP-Star™ or ECF detection reagents.
Specifications for Product Family
Specifikace
Specifikace
  • Catalog No:
  • RPN3688
  • RPN3682
  • RPN3685
  • Aplikace:
  • Sufficient for 5000 cm²
  • Sufficient for 2500 cm²
  • Sufficient for 2500 cm2
Specifikace
Product Family Options
Informace o produktu
  • Sekvence
    Bal.
    Dostupnost
    Price
  • RPN3688
    Amersham AlkPhos Direct Hybridization Buffer
    Amersham AlkPhos Direct Hybridization Buffer
    06
    1
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 23 520,00 Kč
    23 520,00 Kč
    1 * 1 KS
       23 520,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    RPN3688
    Aplikace
    Sufficient for 5000 cm²
  • RPN3682
    Module plus CDP-Star Detection Reagent
    Module plus CDP-Star Detection Reagent
    07
    1
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 17 960,00 Kč
    17 960,00 Kč
    1 * 1 KS
       17 960,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    RPN3682
    Aplikace
    Sufficient for 2500 cm²
  • RPN3685
    ECF Detection Module
    ECF Detection Module
    09
    1
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 21 400,00 Kč
    21 400,00 Kč
    1 * 1 KS
       21 400,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    RPN3685
    Aplikace
    Sufficient for 2500 cm2