Affinity chromatography columns, HiTrap™ Streptavidin HP
Catalog # 17-5112-01
Supplier: Cytiva
Specifications
- Průtoková rychlost:<4 ml/min
- Velikost částic:34 µm
- Maximální tlak:5 bar
- Kapacita:5×1 ml
- Popis:HiTrap™ Streptavidin HP
- Sekvence:01
- Rozsah pH:4 - 9
- Hardware kolony:PP
- Rozměry lůžka:7×25 mm
- Bal.:5
Specifications
About this item
HiTrap Streptavidin HP is a column prepacked with Streptavidin Sepharose High Performance for purification of biotinylated proteins and biomolecules.
- Convenient, fast, reliable purification of biotinylated compounds
- The strong interactions of biotin and streptavidin, as well as the somewhat weaker interactions of 2-iminobiotin and streptavidin are utilised
- The column can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System
Streptavidin Sepharose™ High Performance is also available in lab packs, using the same purified streptavidin isolated from Streptomyces avidinii bound to Sepharose™ High Performance. The prepacked medium ensures reproducibility, high performance, and excellent physical and chemical stability.