Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pufry
Pufry pro elektroforézu
Pufry nanášecí
Native sample loading buffer, 2X
Native sample loading buffer, 2X
Native sample loading buffer, 2X
Catalog # 786-421
Supplier:
G-Biosciences
CAS Number:
Expand image
Native sample loading buffer, 2X
Catalog # 786-421
Supplier:
G-Biosciences
CAS Number:
Specifications
Bal.:
2,5 mL
Popis:
Native sample loading buffer (2X)
Specifications
About this item
Product Details
Product Details