ContactUsLinkComponent
Kjeldahl digestion and distillation assembly, Quickfit®
Kjeldahl digestion and distillation assembly, Quickfit®
Katalogové číslo BIBB280MC
Dodavatel:  SciLabware
undefined
Kjeldahl digestion and distillation assembly, Quickfit®
Katalogové číslo BIBB280MC
Dodavatel:  SciLabware
Specifikace
  • Popis:
    Kjeldhal digestion and distillation unit, with 500 ml capacity Kjeldahl flask
  • Bal.:
    1
  • Typ:
    280MC
Specifikace
O tomto produktu
Convenient assembly of Quickfit® glassware for Kjeldahl digestion (nitrogen determination) and distillation.
  • Manufactured from heat resistant and chemically durable borosilicate glass
Flask can be used alone for digestion stage and then fitted to rest of assembly for the distillation process.