ContactUsLinkComponent
Protein fractionation kit, nuclear and cytoplasmic extraction kit
Protein fractionation kit, nuclear and cytoplasmic extraction kit
  786-182
 :  
undefined
Protein fractionation kit, nuclear and cytoplasmic extraction kit
  786-182
 :  
  • Bal.:
    1 KIT
  • Popis:
    Nuclear and Cytoplasmic Extraction Kit, 100 preps
The Nuclear and Cytoplasmic Extraction kit is for the enrichment of cytoplasmic and nuclear fractions from cultured cells and tissues. The kit generates a cleaner separation of cytoplasmic proteins from nuclear proteins and is ideal for expression and transport studies.
  • For generation of cytosolic and ultra pure nuclear fractions
  • Provided with nuclear protein solubilisation buffers
This kit is based on organic buffers and contains a proprietary combination of various salts and agents. The kit is provided with reagents for solubilisation of nuclear fraction.
The kit is designed for >50 preps, depending on the tissue used, where one prep is 20×10⁶ mammalian cells or 100 mg mammalian tissue.