Orders LinkContactUsLinkComponent
Modular deck system, PIG®
Modular deck system, PIG®
Supplier:   NEW PIG
Modular deck system, PIG®
Supplier:  NEW PIG
Certificates
Modular deck system, PIG®
Modular deck system, PIG®
4 Options Available Below
Product Details & Documents
Modular deck system ideal to create a customised drum storage area that shares sump capacity. With flow-through sump design liquid passes freely from one sump to another.
  • Linear LDPE sump with injection molded PE grate
  • Low-profile design is easy to load or unload
  • Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe
  • LDPE construction resists UV rays, rust, corrosion and most chemicals
  • Textured grating adds traction and lifts out for easy sump access
  • Translucent white sidewalls allow easy visual inspection
Poly loading ramp: Made of lightweight PE. Edge guards guide drum trucks and help prevent them from rolling off the sides. Steel clamps securely attach ramp to pallet or deck.
Steel loading ramp: Heavy dute steel with adjustable legs for use with a range of pallet and deck heights. Edge guards guide drum trucks and help prevent them from rolling off the sides. Lip of ramp secures to edge of pallet or deck. Diamond plate surface provides traction for added safety.
Product Family Options
Poly modular spill deck
Product Information
  • Sekvence
    Popis
    Barva
    Š×H×V
    Hmotnost
    Availability
    Price
  • 01
    Modular spill deck, 42 l sump capacity, 680 kg load capacity, 39% post-industrial recycled polyethylene
    Bílá/černá
    660×660×150 mm
    11,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 950,00 Kč
    4 950,00 Kč
    1 * 1 KS
       4 950,00 Kč
  • 02
    Modular spill deck, 83 l sump capacity, 1360 kg load capacity, 43% post-industrial recycled polyethylene
    Bílá/černá
    1320×660×150 mm
    20,4 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 8 470,00 Kč
    8 470,00 Kč
    1 * 1 KS
       8 470,00 Kč
  • 03
    Modular spill deck, 167 l sump capacity, 2722 kg load capacity, 48% post-industrial recycled polyethylene
    Bílá/černá
    1320×660×150 mm
    34,1 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 12 730,00 Kč
    12 730,00 Kč
    1 * 1 KS
       12 730,00 Kč
Poly loading ramp
Product Information
  • Sekvence
    Popis
    Barva
    Š×H×V
    Hmotnost
    Availability
    Price
  • 04
    Poly loading ramp, 272 kg loading capacity, use with poly spill containment decks 15 cm height
    Černá
    510×810×220 mm
    7,71 kg
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 800,00 Kč
    4 800,00 Kč
    1 * 1 KS
       4 800,00 Kč