Sample tubes for Roche Cobas analysers
2,5 ml, For Roche Cobas analysers, without caps, Bag of 1000, Carton of 2000
Katalogové číslo SIMPB721-1
Dodavatel: Simport Scientific
Přejít na:
Product Details & Documents
PS, clear.
- Good chemical resistance
- Good temperature resistance
This tube is used with the Roche Cobas Analyzer. It has perfect clarity for easy viewing of contents. This graduated false bottom tube has a capacity of 2,5 ml and will accept bar code labels.
Specifikace
- Balené:Bag of 1000, Carton of 2000
- Kapacita:2,5 ml
- Popis:For Roche Cobas analysers, without caps