Injection and aspiration filter, Sterifix® Pury
Catalog # 612-0192
Supplier: B. Braun
Specifications
- Popis:Injection and aspiration filters, 5 μm, with Lock fitting
- Bal.:100
Specifications
About this item
Injection and aspiration filter for the prevention of particulate contamination during injection and aspiration of drugs.