Ion selective electrode system, AmmoLyt® Plus
Catalog # WTWO107042
Supplier: WTW ONLINE
Specifications
- Type:VARiON® Ref
- Popis:Reference electrode, VARiON® Ref, for AmmoLyt® Plus/NitraLyt®Plus 700 IQ/VARiON® Plus
- Bal.:1
Specifications
About this item
This system – consisting of a robust, digital armature with ammonium replacement electrode and ammonium reference electrode – allows ammonium measurement directly in the medium, without sample preparation. The system is designed for connection to the IQ SENSOR NET multiparameter measuring system.
- Calibration-free, long term stable
- With integrated temperature sensor
- Short response time
- With screw/plug connector for connection of the SACIQ sensor connection cable
Ordering information: Supplied with unequipped sensor AmmoLyt® Plus 700 IQ, reference electrode VARiON® Ref, VARiON® Plus NH4 (ammonium electrode), special socket wrench, protective hood VARiON® 700 IQ-SK, Potassium chloride solution for storing the reference electrode and operating manual.