Orders LinkContactUsLinkComponent
398087FeatureCardimg.jpg
Volumetric Solutions, Merck
Titripur® volumetric solutions from Merck.
  • Directly traceable to NIST standard reference materials.
  • Analytical applications in accordance with Reag.Pharm.Eur. and USP, where applicable.
  • Innovative Titripac® packaging - prevents contamination by air, carbon dioxide or microorganisms.  Saves costs and time.
Product selection