Masterflex® Touch Screen Syringe Pump Microliter Rack, for 4 Syringes
Catalog # MFLB74905-62
Supplier: MASTERFLEX PREMIUM
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
- Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
- 说明:Syringe Pump Microliter Rack
- Environmentally Preferable:
- 型号:74905-62
- Syringe size accepted:0,5 µl - 10 ml
- 质保:1 year
- 包装规格:1
- No. of syringes:4