Masterflex® Fitting, Brass, Straight, Hosebarb to Thread Adapter, 1/4" ID x 1/8" NPT(M); 50/PK
目录编号 MFLB30905-01
供应商: MASTERFLEX PREMIUM
关于此项目
安装快速简便。
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/4" ID x 1/8" NPT(M)
多种材质和配置,满足您的需求。NPT阳螺纹软管倒刺接头。
- Barb size/tubing int.Ø (port 2):¼"
- Connection Type:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):⅛"
- 说明:Fitting, Brass, Straight, Hosebarb to Thread Adapter, ¼" ID × ⅛" NPT(M); 50/PK
- 顺序:19
- Environmentally Preferable:
- 型号:30905-01
- No. of ports:2
- 质保:1 year
- 包装规格:50
- Fitting material:Brass