Micropump L24566 Pump Head Adapter Kit; NEMA 56C to A-Mount, High-Torque Mag
目录编号 MFLB07003-96
供应商: MASTERFLEX PREMIUM
关于此项目
Easily adapt your pump head to your drive with these adapter kits.
- Choose adapter kit suitable for use with your Masterflex pump head
使用此适配器套件,可将微型泵的A形安装架泵头连接到您的电动机或Masterflex®L/S®泵驱动器上。
- 说明:Pump Head Adapter Kit; NEMA 56C to A-Mount, High-Torque Mag
- Environmentally Preferable:
- 包装规格:1