Masterflex® Dual-Drive Syringe Pump RS-232 Connector Cable
目录编号 MFLB74901-88
供应商: MASTERFLEX PREMIUM
关于此项目
Daisy chain up to 100 pumps.
- Connect multiple independent syringes to operate simultaneously
- Easily connect pumps
- Choose cable based on your current pump
Optimize performance of your Masterflex® Dual-Drive Syringe Pump with these accessories.
- 说明:Pumps, Syringe Pumps RS-232 Connector Cable for 74901-80 Dual Drive Syringe Pump
- 顺序:02
- Environmentally Preferable:
- 型号:74901-88
- Outlet connection:RS-232 Connector Cable for 74901-80
- 质保:1 year
- 包装规格:1